Läs mer
1,3,5-Tris(4-aminophenyl)benzene, TRC
Produktkod. 30240547
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240547 500 mg
30240839 50 mg
30240694 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT875008500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 118727-34-7
Molekylformel C24H21N3
Molekylvikt (g/mol) 351.44
IUPAC-namn 4-[3,5-bis(4-aminophenyl)phenyl]aniline
LEDER Nc1ccc(cc1)c2cc(cc(c2)c3ccc(N)cc3)c4ccc(N)cc4
Specifikationer
Kemiskt namn eller material 1,3,5-Tris(4-aminophenyl)benzene
Kvantitet 500 mg
InChI formel InChI=1S/C24H21N3/c25-22-7-1-16(2-8-22)19-13-20(17-3-9-23(26)10-4-17)15-21(14-19)18-5-11-24(27)12-6-18/h1-15H,25-27H2
Formel vikt 351.1735