Learn More
1,3,6,8-Tetrabromopyrene, TRC
Product Code. 30422003
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30422003 5 g 5g
1 options
Product Code. 30422003 Supplier Toronto Research Chemicals Supplier No. TRCT2911855G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3,6,8-Tetrabromopyrene
CAS 128-63-2
Molecular Formula C16H6Br4
Quantity 5 g
Synonym Pyrene','1,3,6,8-tetrabromo- (6CI','8CI','9CI','ACI), 1,3,6,8-Tetrabromopyrene (ACI), Pyrene','1,3,6,8-tetrabromo-1,9-dihydro-
InChI Formula InChI=1S/C16H6Br4/c17-11-5-13(19)9-3-4-10-14(20)6-12(18)8-2-1-7(11)15(9)16(8)10/h1-6H
SMILES C1=CC2=C3C(=C(C=C2Br)Br)C=CC4=C(C=C(C1=C43)Br)Br
IUPAC Name 1,3,6,8-tetrabromopyrene
Molecular Weight (g/mol) 517.83
Formula Weight 513.7203