missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-5-glycolic Acid, TRC

Product Code. 30334242
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30334242 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334242 Supplier Toronto Research Chemicals Supplier No. TRCB410518100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1,3-Benzodioxole-5-glycolic Acid
CAS 27738-46-1
Molekylformel C9H8O5
Kvantitet 100 mg
Synonym alpha-Hydroxy-1,3-benzodioxole-5-acetic Acid, 2-(1,3-Benzodioxol-5-yl)-2-hydroxyacetic Acid, 2-(2H-1,3-Benzodioxol-5-yl)-2-hydroxyacetic Acid, 2-Hydroxy-2-(3,4-methylenedioxyphenyl)acetic Acid, 3,4-(Methylenedioxy)benzeneglycolic Acid, 3,4-(Methylenedioxy)mandelic Acid, 3',4'-Methylenedioxymandelic Acid
InChI formel InChI=1S/C9H8O5/c10-8(9(11)12)5-1-2-6-7(3-5)14-4-13-6/h1-3,8,10H,4H2,(H,11,12)
LEDER OC(C(=O)O)c1ccc2OCOc2c1
IUPAC-namn 2-(1,3-benzodioxol-5-yl)-2-hydroxyacetic acid
Molekylvikt (g/mol) 196.16
Formel vikt 196.0372
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.