Läs mer
1,3-Bis(1-naphthalenyloxy)-2-propanol, TRC
Produktkod. 30004326
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004326 5 g
17922105 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004326 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5043005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17216-10-3
Molekylformel C23 H20 O3
Molekylvikt (g/mol) 344.4
Synonym 1,3-Bis(naphthalen-1-yloxy)-propan-2-ol, Propranolol Hydrochloride Imp. C (EP), Bis-ether Derivative of Propranolol
IUPAC-namn 1,3-dinaphthalen-1-yloxypropan-2-ol
LEDER OC(COc1cccc2ccccc12)COc3cccc4ccccc34
Specifikationer
Kemiskt namn eller material 1,3-Bis(naphthalen-1-yloxy)-propan-2-ol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C23H20O3/c24-19(15-25-22-13-5-9-17-7-1-3-11-20(17)22)16-26-23-14-6-10-18-8-2-4-12-21(18)23/h1-14,19,24H,15-16H2
Formel vikt 344.1412
Procent renhet >95%