Läs mer
1,3-Bis(2-isocyanato-2-propyl)benzene, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,3-Bis(2-isocyanato-2-propyl)benzene |
| CAS | 2778-42-9 |
| Molekylformel | C14 H16 N2 O2 |
| Kvantitet | 2.5 g |
| Synonym | Isocyanic acid, m-phenylenediisopropylidene ester (7CI), Isocyanic acid, α,α,α',α'-tetramethyl-m-xylylene ester (8CI), 1,3-Bis(1-isocyanato-1-methylethyl)benzene, 1,3-Bis(1-isocyanatolmethylethyl)benzene, 1,3-Bis(1-methyl-1-isocyanatoethyl)benzene, 1,3-Bis(2-isocyanato-prop-2-yl)benzene, 1,3-Bis(2-isocyanatopropan-2-yl)benzene, 1,3-Bis(α-isocyanatoisopropyl)benzene, Tetramethyl-1,3-xylylene diisocyanate, Tetramethyl-m-xylylene diisocyanate, m-Bis(1-isocyanato-1-methylethyl)benzene, m-TMXDI, m-Tetramethylxylene diisocyanate, α,α,α',α'-Tetramethyl-1,3-xylylen diisocyanate, α,α,α',α'-Tetramethyl-1,3-xylylene diisocyanate, α,α,α',α'-Tetramethyl-m-phenylenedimethylene diisocyanate, α,α,α',α'-Tetramethyl-m-xylene diisocyanate, α,α,α',α'-Tetramethyl-m-xylylene diisocyanate |
| InChI formel | InChI=1S/C14H16N2O2/c1-13(2,15-9-17)11-6-5-7-12(8-11)14(3,4)16-10-18/h5-8H,1-4H3 |
| LEDER | CC(C)(N=C=O)c1cccc(c1)C(C)(C)N=C=O |
| IUPAC-namn | 1,3-bis(2-isocyanatopropan-2-yl)benzene |
| Molekylvikt (g/mol) | 244.29 |
| Formel vikt | 244.1212 |