missing translation for 'onlineSavingsMsg'
Läs mer

1,3-Bis(2-methoxyphenoxy)-2-propanol, TRC

Produktkod. 30331586
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30331586 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30331586 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB49003050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1,3-Bis(2-methoxyphenoxy)propan-2-ol
CAS 16929-60-5
Anteckningar om renhetsgrad HPLC
Molekylformel C17 H20 O5
Kvantitet 50 mg
Synonym 1,3-Bis(2-methoxyphenoxy)propan-2-ol, Guaifenesin Imp. D (EP)
InChI formel InChI=1S/C17H20O5/c1-19-14-7-3-5-9-16(14)21-11-13(18)12-22-17-10-6-4-8-15(17)20-2/h3-10,13,18H,11-12H2,1-2H3
LEDER COc1ccccc1OCC(O)COc2ccccc2OC
IUPAC-namn 1,3-bis(2-methoxyphenoxy)propan-2-ol
Molekylvikt (g/mol) 304.34
Formel vikt 304.1311
Procent renhet >95
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.