Learn More
1,3-Bis(2-propynyloxy)benzene, 97%, BLD Pharm™
Product Code. 30945243
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30945243 250 mg 250mg
30935321 1 g 1g
30929699 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30945243 Supplier BLD Pharm Supplier No. BD00797469250mg
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 26627-36-1
Molecular Formula C12H10O2
MDL Number MFCD00224312
Quantity 250 mg
SMILES C#CCOC1=CC(OCC#C)=CC=C1
Molecular Weight (g/mol) 186.21
Percent Purity 97%
Chemical Name or Material 1,3-Bis(2-propynyloxy)benzene