Läs mer
1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane, TRC
Produktkod. 30138023
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138023 500 mg
30008158 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB419055500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6323-09-7
Molekylformel C23H30Cl2N4
Molekylvikt (g/mol) 433.42
Synonym 1,1'-(1,3-Propanediyl)bis[4-(3-chlorophenyl)piperazine, 1,1'-Trimethylenebis[4-(m-chlorophenyl)piperazine, NSC 32568, Trazodone Impurity H, Bispiperazine Trazodone
IUPAC-namn (1-hydroxy-2-methylpropan-2-yl) acetate
LEDER CC(=O)OC(C)(C)CO
Specifikationer
Kemiskt namn eller material 1,3-Bis-[4-(3-chlorophenyl)piperazin-1-yl]propane
Kvantitet 500 mg
InChI formel InChI=1S/C6H12O3/c1-5(8)9-6(2,3)4-7/h7H,4H2,1-3H3