Learn More
Alfa Aesar™ 1-[3-(Boc-amino)propyl]indole-6-carboxylic acid, 97%
Brand:  Alfa Aesar™ H63083.03
Additional Details : CAS Number : 1005378-70-0 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15488116
  • 4955.73 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
1-[3-(Boc-amino)propyl]indole-6-carboxylic acid
1005378-70-0
318.37
MFCD04973975
Specifications
1-[3-(Boc-amino)propyl]indole-6-carboxylic acid
Solid
318.37
1g
1005378-70-0
97%
MFCD04973975