Läs mer
1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine, TRC
Produktkod. 30133535
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133535 25 mg -
30006540 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17905525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61338-13-4
Molekylformel C17 H19 N3 O2
Molekylvikt (g/mol) 297.35
Synonym 2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinecarboxylic acid, 1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine
IUPAC-namn 2-(4-methyl-2-phenylpiperazin-1-yl)pyridine-3-carboxylic acid
LEDER CN1CCN(C(C1)c2ccccc2)c3ncccc3C(=O)O
Specifikationer
Kemiskt namn eller material 1-(3-Carboxy-2-pyridyl)-4-methyl-2-phenylpiperazine
Kvantitet 25 mg
InChI formel InChI=1S/C17H19N3O2/c1-19-10-11-20(15(12-19)13-6-3-2-4-7-13)16-14(17(21)22)8-5-9-18-16/h2-9,15H,10-12H2,1H3,(H,21,22)
Formel vikt 297.1477