Läs mer
1-(3-Carboxypropyl)-3,7-dimethylxanthine, TRC
Produktkod. 30138467
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138467 50 mg
30006343 10 mg
30007506 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18123550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6493-07-8
Molekylformel C11 H14 N4 O4
Molekylvikt (g/mol) 266.25
Synonym 1H-Purine-1-butanoic acid, 2,3,6,7-tetrahydro-3,7-dimethyl-2,6-dioxo-, Purine-1(2H)-butyric acid, 3,6-dihydro-3,7-dimethyl-2,6-dioxo- (7CI,8CI), 2,3,6,7-Tetrahydro-3,7-dimethyl-2,6-dioxo-1H-purine-1-butanoic acid, 1-(3-Carboxypropyl)-3,7-dimethylxanthine, 1-(3-Carboxypropyl)3,7-dimethylxantine, 1-(3-Carboxypropyl)theobromine, 1-(3'-Carboxypropyl)-3,7-dimethylxanthine, 3,7-Dimethyl-1-(3-carboxypropyl)xanthine, C 21187
IUPAC-namn 4-(3,7-dimethyl-2,6-dioxopurin-1-yl)butanoic acid
LEDER CN1C(=O)N(CCCC(=O)O)C(=O)c2c1ncn2C
Specifikationer
Kemiskt namn eller material 1-(3-Carboxypropyl)-3,7-dimethylxanthine
Kvantitet 50 mg
InChI formel InChI=1S/C11H14N4O4/c1-13-6-12-9-8(13)10(18)15(11(19)14(9)2)5-3-4-7(16)17/h6H,3-5H2,1-2H3,(H,16,17)
Formel vikt 266.1015