Läs mer
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine Hydrochloride, TRC
Produktkod. 30351997
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30351997 10 g
1 options
Produktkod. 30351997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37772010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride
CAS 52605-52-4
Molekylformel C13 H18 Cl2 N2 . Cl H
Kvantitet 10 g
Synonym 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride, Trazodone Hydrochloride Imp. F (BP) as Hydrochloride, Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, hydrochloride (1:1), Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, monohydrochloride (9CI), 1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride (1:1)
InChI formel InChI=1S/C13H18Cl2N2.ClH/c14-5-2-6-16-7-9-17(10-8-16)13-4-1-3-12(15)11-13;/h1,3-4,11H,2,5-10H2;1H
LEDER Cl.ClCCCN1CCN(CC1)c2cccc(Cl)c2
IUPAC-namn 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazine;hydrochloride
Molekylvikt (g/mol) 309.66
Formel vikt 308.0614