Läs mer
1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride, TRC
Produktkod. 30366467
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30366467 5 g
1 options
Produktkod. 30366467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3797455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride
CAS 2031-23-4
Molekylformel C8H19Cl3N2
Kvantitet 5 g
Synonym 1-(3-Chloropropyl)-4-methyl-piperazine Hydrochloride (1:2), 1-Chloro-3-(4-methyl-1-piperazinyl)propane Dihydrochloride, 1-Methyl-4-(3-chloropropyl)piperazine Dihydrochloride, 3-(4-Methylpiperazino)propyl Chloride Dihydrochloride, N-(3-Chloropropyl)-N'-methylpiperazine Dihydrochloride
InChI formel InChI=1S/C8H17ClN2.2ClH/c1-10-5-7-11(8-6-10)4-2-3-9;;/h2-8H2,1H3;2*1H
LEDER Cl.Cl.CN1CCN(CCCCl)CC1
IUPAC-namn 1-(3-chloropropyl)-4-methylpiperazine;dihydrochloride
Molekylvikt (g/mol) 249.61