missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride, TRC

Product Code. 30353132
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30353132 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353132 Supplier Toronto Research Chemicals Supplier No. TRCC3797451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride
CAS 2031-23-4
Molekylformel C8H19Cl3N2
Kvantitet 1 g
Synonym 1-(3-Chloropropyl)-4-methyl-piperazine Hydrochloride (1:2), 1-Chloro-3-(4-methyl-1-piperazinyl)propane Dihydrochloride, 1-Methyl-4-(3-chloropropyl)piperazine Dihydrochloride, 3-(4-Methylpiperazino)propyl Chloride Dihydrochloride, N-(3-Chloropropyl)-N'-methylpiperazine Dihydrochloride
InChI formel InChI=1S/C8H17ClN2.2ClH/c1-10-5-7-11(8-6-10)4-2-3-9;;/h2-8H2,1H3;2*1H
LEDER Cl.Cl.CN1CCN(CCCCl)CC1
IUPAC-namn 1-(3-chloropropyl)-4-methylpiperazine;dihydrochloride
Molekylvikt (g/mol) 249.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.