Läs mer
1,3-Diacetoxypropane, TRC
Produktkod. 30354681
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354681 2.5 g
30348818 500 mg
30363115 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354681 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3075732.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 628-66-0
Molekylformel C7H12O4
Molekylvikt (g/mol) 160.17
IUPAC-namn 3-acetyloxypropyl acetate
LEDER CC(=O)OCCCOC(=O)C
Specifikationer
Kemiskt namn eller material 1,3-Diacetoxypropane
Kvantitet 2.5 g
InChI formel InChI=1S/C7H12O4/c1-6(8)10-4-3-5-11-7(2)9/h3-5H2,1-2H3
Formel vikt 160.0736