Läs mer
(1,3-Dichloropropyl)benzene, TRC
Produktkod. 30368485
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368485 1 g
30366332 500 mg
30353058 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1884701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14155-36-3
Molekylformel C9H10Cl2
Molekylvikt (g/mol) 189.08
Synonym 1,3-Dichloro-1-phenylpropane
IUPAC-namn 1,3-dichloropropylbenzene
LEDER ClCCC(Cl)c1ccccc1
Specifikationer
Kemiskt namn eller material (1,3-Dichloropropyl)benzene
Kvantitet 1 g
InChI formel InChI=1S/C9H10Cl2/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9H,6-7H2
Formel vikt 188.016