Läs mer
1,3-Diethyl-6-aminouracil, TRC
Produktkod. 30377911
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30377911 250 mg
30369084 1 g
30373867 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30377911 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD443675250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41740-15-2
Molekylformel C8 H13 N3 O2
Molekylvikt (g/mol) 183.21
IUPAC-namn 6-amino-1,3-diethylpyrimidine-2,4-dione
LEDER CCN1C(=CC(=O)N(CC)C1=O)N
Specifikationer
Kemiskt namn eller material 1,3-Diethyl-6-aminouracil
Kvantitet 250 mg
InChI formel InChI=1S/C8H13N3O2/c1-3-10-6(9)5-7(12)11(4-2)8(10)13/h5H,3-4,9H2,1-2H3
Formel vikt 183.1008