Läs mer
1,3-Difluoropropane, 97%, Thermo Scientific™
Produktkod. 11388755
Change view
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11388755 25 g 25g
11378755 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11388755 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L12351.14
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 462-39-5
Molekylformel C3H6F2
Molekylvikt (g/mol) 80.08
MDL-nummer MFCD00040969
InChI-nyckel OOLOYCGJRJFTPM-UHFFFAOYSA-N
Synonym propane, 1,3-difluoro, acmc-1aiey, 1,3-bis fluoranyl propane
PubChem CID 68041
IUPAC-namn 1,3-difluoropropane
LEDER FCCCF
Specifikationer
Kemiskt namn eller material 1,3-Difluoropropane
Densitet 1.006
Kokpunkt 40°C to 42°C
Linjär formel FCH2CH2CH2F
Brytningsindex 1.321
Kvantitet 25 g
Beilstein 1696999
Formel vikt 80.08
Procent renhet 97%