Läs mer
1,3-Dihydro-2-oxo-4-(4-methyl-1-piperazinyl)-3H-1,5-benzodiazepine, TRC
Produktkod. 30379301
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379301 250 mg
30374936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379301 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453430250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,3-Dihydro-2-oxo-4-(4-methyl-1-piperazinyl)-3H-1,5-benzodiazepine
CAS 1391053-73-8
Molekylformel C14H18N4O
Kvantitet 250 mg
InChI formel InChI=1S/C14H18N4O/c1-17-6-8-18(9-7-17)13-10-14(19)16-12-5-3-2-4-11(12)15-13/h2-5H,6-10H2,1H3,(H,16,19)
LEDER CN1CCN(CC1)C2=Nc3ccccc3NC(=O)C2
IUPAC-namn 4-(4-methylpiperazin-1-yl)-1,3-dihydro-1,5-benzodiazepin-2-one
Molekylvikt (g/mol) 258.32
Formel vikt 258.1481