missing translation for 'onlineSavingsMsg'
Läs mer

1,3-Dihydro-N-nitro-1,3-dioxo-2H-isoindole-2-carboximidothioic acid Methyl Ester, TRC

Produktkod. 30377202
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30377202 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30377202 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453990500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1,3-Dihydro-N-nitro-1,3-dioxo-2H-isoindole-2-carboximidothioic acid Methyl Ester
CAS 138149-96-9
Molekylformel C10H7N3O4S
Kvantitet 500 mg
InChI formel InChI=1S/C10H7N3O4S/c1-18-10(11-13(16)17)12-8(14)6-4-2-3-5-7(6)9(12)15/h2-5H,1H3
LEDER O=C1C=2C=CC=CC2C(=O)N1C(=NN(=O)=O)SC
IUPAC-namn methyl N-nitro-1,3-dioxoisoindoline-2-carbimidothioate
Molekylvikt (g/mol) 265.25
Formel vikt 265.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.