missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7047042
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
626-00-6
C6H4I2
329.907
MFCD00041731
SFPQFQUXAJOWNF-UHFFFAOYSA-N
m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
12270
1,3-diiodobenzene
C1=CC(=CC(=C1)I)I
This item is not returnable. View return policy
626-00-6
C6H4I2
329.907
MFCD00041731
SFPQFQUXAJOWNF-UHFFFAOYSA-N
m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
12270
1,3-diiodobenzene
C1=CC(=CC(=C1)I)I

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in hot methanol.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 35°C to 37°C
Density 2.47
Boiling Point 285°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1904540
Sensitivity Light sensitive
Solubility Information Insoluble in water. Soluble in hot methanol.
Formula Weight 329.91
Percent Purity ≥98%
Chemical Name or Material 1,3-Diiodobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'