missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7021156
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
151-10-0
C8H10O2
138.17
MFCD00008384
DPZNOMCNRMUKPS-UHFFFAOYSA-N
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
9025
1,3-dimethoxybenzene
COC1=CC(OC)=CC=C1
This item is not returnable. View return policy
151-10-0
C8H10O2
138.17
MFCD00008384
DPZNOMCNRMUKPS-UHFFFAOYSA-N
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
9025
1,3-dimethoxybenzene
COC1=CC(OC)=CC=C1

1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -52°C
Density 1.065
Boiling Point 217°C to 218°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.525
Quantity 100 g
Beilstein 878582
Solubility Information Miscible with toluene.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,3-Dimethoxybenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'