Läs mer
1,3-Dimethylpentylamine Hydrochloride Salt, TRC
Produktkod. 30132430
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132430 500 mg
30132567 50 mg
17984115 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD476600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13803-74-2
Molekylformel C7 H17 N . Cl H
Molekylvikt (g/mol) 151.68
Synonym Dimethylamylamine Hydrochloride, 4-Methyl-2-hexanamine Hydrochloride, DMAA HCl
IUPAC-namn 4-methylhexan-2-amine;hydrochloride
LEDER Cl.CCC(C)CC(C)N
Specifikationer
Kemiskt namn eller material Methylhexanamine hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C7H17N.ClH/c1-4-6(2)5-7(3)8;/h6-7H,4-5,8H2,1-3H3;1H
Formel vikt 151.1128