Läs mer
1,3-Dinitrobenzene, TRC
Produktkod. 30137789
Change view
Kvantitet:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137789 250 g
30138231 50 g
18080200 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137789 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD479835250G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99-65-0
Molekylformel C6 H4 N2 O4
Molekylvikt (g/mol) 168.11
Synonym Benzene, 1,3-dinitro-, Benzene, m-dinitro- (8CI), 1,3-Dinitrobenzene, DNB, DNB (explosive), Meta-dinitrobenzene, NSC 7189, m-Dinitrobenzene, m-Dinitrobenzene
IUPAC-namn 1,3-dinitrobenzene
LEDER [O-][N+](=O)c1cccc(c1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 1,3-Dinitrobenzene
Kvantitet 250 g
InChI formel InChI=1S/C6H4N2O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H
Formel vikt 168.0171