missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1,3-Dioctanoyl Glycerol, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,3-Dioctanoyl Glycerol |
| CAS | 1429-66-9 |
| Molekylformel | C19H36O5 |
| Kvantitet | 500 mg |
| Synonym | Glycerol 1,3-Dicaprylate, 1,3-Dioctanoyl Glycerol, 1,3-Dioctanoin, Octanoic Acid 2-Hydoxy-1,3-propanediyl Ester, Octanoic Acid, 1,1'-(2-Hydroxy-1,3-propanediyl) Ester |
| InChI formel | InChI=1S/C19H36O5/c1-3-5-7-9-11-13-18(21)23-15-17(20)16-24-19(22)14-12-10-8-6-4-2/h17,20H,3-16H2,1-2H3 |
| LEDER | CC(C)=CC1OC(=O)C=C(C)C=1 |
| Molekylvikt (g/mol) | 344.49 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?