Läs mer
1,3-Dioctanoyl Glycerol, TRC
Produktkod. 30373961
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373961 100 mg
1 options
Produktkod. 30373961 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD481010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,3-Dioctanoyl Glycerol
CAS 1429-66-9
Molekylformel C19H36O5
Kvantitet 100 mg
Synonym Glycerol 1,3-Dicaprylate, 1,3-Dioctanoyl Glycerol, 1,3-Dioctanoin, Octanoic Acid 2-Hydoxy-1,3-propanediyl Ester, Octanoic Acid, 1,1'-(2-Hydroxy-1,3-propanediyl) Ester
InChI formel InChI=1S/C19H36O5/c1-3-5-7-9-11-13-18(21)23-15-17(20)16-24-19(22)14-12-10-8-6-4-2/h17,20H,3-16H2,1-2H3
LEDER CC(C)=CC1OC(=O)C=C(C)C=1
Molekylvikt (g/mol) 344.49