Läs mer
1,3-Dioleoyl-2-palmitoylglycerol, TRC
Produktkod. 30011650
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011650 25 mg
30011542 10 mg
30009451 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48420525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1716-07-0
Molekylformel C55 H102 O6
Molekylvikt (g/mol) 859.39
IUPAC-namn [2-hexadecanoyloxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
LEDER CCCCCCCCCCCCCCCC(=O)OC(COC(=O)CCCCCCC\C=C\CCCCCCCC)COC(=O)CCCCCCC\C=C\CCCCCCCC
Specifikationer
Kemiskt namn eller material 1,3-Dioleoyl-2-palmitoylglycerol
Kvantitet 25 mg
InChI formel InChI=1S/C55H102O6/c1-4-7-10-13-16-19-22-25-27-30-32-35-38-41-44-47-53(56)59-50-52(61-55(58)49-46-43-40-37-34-29-24-21-18-15-12-9-6-3)51-60-54(57)48-45-42-39-36-33-31-28-26-23-20-17-14-11-8-5-2/h25-28,52H,4-24,29-51H2,1-3H3/b27-25+,28-26+
Formel vikt 858.7676