missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.8%, anhydrous, stabilized with 75 ppm BHT, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229191
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
646-06-0, 128-37-0
C3H6O2
74.08
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
31404
CHEBI:34247
2,6-ditert-butyl-4-methylphenol
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
This item is not returnable. View return policy
646-06-0, 128-37-0
C3H6O2
74.08
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
31404
CHEBI:34247
2,6-ditert-butyl-4-methylphenol
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.75
CAS Max % 100.0
Melting Point -95°C
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Ignition Residue 0.0005% max
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3990 to 1.4010
Quantity 100 mL
Residue after Evaporation 0.0005% max.
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm (BHT)
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.8%
Physical Form Liquid
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dioxolane, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'