Läs mer
1,3-Dipalmitoyl-2-chloropropanediol, TRC
Produktkod. 17969854
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17969854 25 mg
17968124 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17969854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48684025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 169471-41-4
Molekylformel C35 H67 Cl O4
Molekylvikt (g/mol) 587.36
Synonym Hexadecanoic acid, 1,1'-(2-chloro-1,3-propanediyl) ester, Hexadecanoic acid, 2-chloro-1,3-propanediyl ester (9CI), 1,1'-(2-Chloro-1,3-propanediyl) dihexadecanoate, 1,2-Dipalmitoyl-3-chloropropane
IUPAC-namn (2-chloro-3-hexadecanoyloxypropyl) hexadecanoate
LEDER CCCCCCCCCCCCCCCC(=O)OCC(Cl)COC(=O)CCCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material 1,3-Dipalmitoyl-2-chloropropanediol
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C35H67ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3
Formel vikt 586.4728
Procent renhet >95%