Läs mer
1,3-Dipalmitoyl-2-oleoyl Glycerol, TRC
Produktkod. 30382015
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30382015 500 mg
30378143 25 mg
30380218 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30382015 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD486845500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2190-25-2
Molekylformel C53H100O6
Molekylvikt (g/mol) 833.36
Synonym (9Z)-9-Octadecenoic Acid 2-[(1-Oxohexadecyl)oxy]-1-[[(1-oxohexadecyl)oxy]methyl]ethyl Ester, 1,3-Dipalmito-2-olein, 1,3-Dipalmitoyl-2-olein, 1,3-Dipalmitoyl-2-oleoylglycerin, 1,3-Dipalmitoyl-2-oleoylglycerol, 1-Palmito-2-oleo-3-palmitin, 2-Oleo-1,3-dipalmitin, 2-Oleodipalmitin, 2-Oleoyl-1,3-dipalmitin, 2-Oleoyl-1,3-dipalmitoylglycerin, 2-Oleoyl-1,3-dipalmitoylglycerol, Glycerin-1-palmitate-2-oleate-3-palmitate, Glycerol 1,3-Dipalmitate 2-oleate, Glyceryl 2-oleate-1,3-dipalmitate, POP
LEDER CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC
Specifications
Kemiskt namn eller material 1,3-Dipalmitoyl-2-oleoyl Glycerol
Kvantitet 500 mg
InChI formel InChI=1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25-
Formel vikt 832.752