Läs mer
1,3-Dithiolan-2-imine, TRC
Produktkod. 30377018
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30377018 250 mg
1 options
Produktkod. 30377018 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD494695250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,3-Dithiolan-2-imine
CAS 4472-81-5
Molekylformel C3H5NS2
Kvantitet 250 mg
Synonym Dithioimidocarbonic Acid Cyclic Ethylene Ester, 1,2-Ethanedithiol Cyclic Dithioimidocarbonate, 1,2-Ethanedithiol Cyclic Ester With Dithioimidocarbonic Acid, Carbonimidodithioic Acid Cyclic 1,2-Ethanediyl Ester
InChI formel InChI=1S/C3H5NS2/c4-3-5-1-2-6-3/h4H,1-2H2
LEDER N=C1SCCS1
IUPAC-namn 1,3-dithiolan-2-imine
Molekylvikt (g/mol) 119.21
Formel vikt 118.9863