Learn More
Alfa Aesar™ 1-(3-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand:  Alfa Aesar™ H64564.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15458986
  • 7780.68 SEK / 5g
Estimated Shipment date
from Supplier 06-07-2018
Add to basket
Specifications
Chemical Identifiers
1-(3-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
328.26
MFCD18185871
Specifications
1-(3-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
328.26
5g
97%
MFCD18185871