Läs mer
1-(3-Methoxyphenyl)ethanol, TRC
Produktkod. 30478714
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30478714 50 mg
1 options
Produktkod. 30478714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32960050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(3-Methoxyphenyl)ethanol
CAS 23308-82-9
Molekylformel C9 H12 O2
Kvantitet 50 mg
Synonym Benzenemethanol, 3-methoxy-α-methyl-, Benzyl alcohol, m-methoxy-α-methyl- (6CI,8CI), 3-Methoxy-α-methylbenzenemethanol, (±)-1-(3-Methoxyphenyl)ethanol, 1-(1-Hydroxyethyl)-3-methoxybenzene, 1-(3-Methoxyphenyl)ethanol, 1-(3'-Methoxyphenyl)ethanol, 3-Methoxy-α-methylbenzyl alcohol, NSC 143364, α-(3-Methoxyphenyl)ethanol, α-(m-Methoxyphenyl)ethanol, Rivastigmine Hydrogen Tartrate Imp. G (EP)
InChI formel InChI=1S/C9H12O2/c1-7(10)8-4-3-5-9(6-8)11-2/h3-7,10H,1-2H3
LEDER COc1cccc(c1)C(C)O
IUPAC-namn 1-(3-methoxyphenyl)ethanol
Molekylvikt (g/mol) 152.19
Formel vikt 152.0837