Läs mer
1-(3-Methoxyphenyl)piperazine dihydrochloride, 97%, Thermo Scientific™
Produktkod. 11301537
Change view
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11301537 5 g 5g
11311537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11301537 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L02164.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-(3-Methoxyphenyl)piperazine dihydrochloride is used as an arylpiperazine compound which is an important intermediate in the preparation of various pharmaceuticals, used to treat depression and post-traumatic stress.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6968-76-9
Molekylformel C11H18Cl2N2O
Molekylvikt (g/mol) 265.178
MDL-nummer MFCD00035285
InChI-nyckel UKUNKQNESKRETR-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine dihydrochloride, unii-q0t82qhg7n, 1-3-methoxyphenyl piperazine 2hcl, q0t82qhg7n, piperazine, 1-3-methoxyphenyl-, dihydrochloride, 1-3-methoxyphenyl piperazindihydrochlorid, 1-3-methoxyphenyl piperazine hcl, pubchem15280, 1-3-methoxyphenyl piperazinehydrochloride, n-3-methoxyphenyl piperazine dihydrochloride
PubChem CID 81429
IUPAC-namn 1-(3-methoxyphenyl)piperazine;dihydrochloride
LEDER COC1=CC=CC(=C1)N2CCNCC2.Cl.Cl
Specifikationer
Kemiskt namn eller material 1-(3-Methoxyphenyl)piperazine dihydrochloride
Smältpunkt ∽214°C (decomposition)
Kvantitet 5 g
Beilstein 5647713
Känslighet Hygroscopic
Löslighetsinformation Soluble in water.
Formel vikt 265.18
Procent renhet 97%