missing translation for 'onlineSavingsMsg'
Läs mer

1-(3-Methyl-1-oxido-2-quinoxalinyl)-ethanone, TRC

Produktkod. 30467716
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30467716 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467716 Supplier Toronto Research Chemicals Supplier No. TRCM312445100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-(3-Methyl-1-oxido-2-quinoxalinyl)-ethanone
CAS 61522-57-4
Molekylformel C11 H10 N2 O2
Kvantitet 100 mg
Synonym 1-(3-Methyl-1-oxido-2-quinoxalinyl)ethanone, 2-Acetyl-3-methylquinoxaline 1-oxide, 1-(3-methyl-2-quinoxalinyl)-N1-oxide Ethanone, 2-Acetyl-3-methylquinoxaline 1-oxide
InChI formel InChI=1S/C11H10N2O2/c1-7-11(8(2)14)13(15)10-6-4-3-5-9(10)12-7/h3-6H,1-2H3
LEDER CC(=O)c1c(C)nc2ccccc2n1=O
IUPAC-namn 1-(3-methyl-1-oxoquinoxalin-2-yl)ethanone
Molekylvikt (g/mol) 202.209
Formel vikt 202.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.