Läs mer
1-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)piperazine, TRC
Produktkod. 30478798
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478798 500 mg
30471539 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30478798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM324795500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 401566-79-8
Molekylformel C14H18N4
Molekylvikt (g/mol) 242.32
Synonym 1-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)-piperazine
IUPAC-namn 1-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazine
LEDER CC1=NN(C2=CC=CC=C2)C(N3CCNCC3)=C1
Specifikationer
Kemiskt namn eller material 1-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)piperazine
Kvantitet 500 mg
InChI formel InChI=1S/C14H18N4/c1-12-11-14(17-9-7-15-8-10-17)18(16-12)13-5-3-2-4-6-13/h2-6,11,15H,7-10H2,1H3
Formel vikt 242.15