Learn More
Alfa Aesar™ 1-(3-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%
Brand:  Alfa Aesar™ H61577.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15476616
  • 5659.56 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
1-(3-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea
308.3
MFCD26143291
Specifications
1-(3-Methylbenzyl)-3-[4-(trifluoromethyl)phenyl]urea
97%
MFCD26143291
1g
Solid
308.3