missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methylphenyl)piperazine, 99%, Thermo Scientific™

Catalog Number p-166258
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
41186-03-2
C11H16N2
176.26
MFCD00040734
JIWHIRLNKIUYSM-UHFFFAOYSA-N
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine
83111
1-(3-methylphenyl)piperazine
CC1=CC(=CC=C1)N2CCNCC2
This item is not returnable. View return policy
41186-03-2
C11H16N2
176.26
MFCD00040734
JIWHIRLNKIUYSM-UHFFFAOYSA-N
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine
83111
1-(3-methylphenyl)piperazine
CC1=CC(=CC=C1)N2CCNCC2
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5755 to 1.5785
Quantity 1 g
Formula Weight 176.26
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 1-(3-Methylphenyl)piperazine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(3-Methylphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'