Läs mer
1,3-O-Bis(triisopropylsilyl) Glycerol, TRC
Produktkod. 30420564
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420564 25 g
30412717 5 g
30409344 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29635025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98264-26-7
Molekylformel C21H48O3Si2
Molekylvikt (g/mol) 404.77
Synonym 3,3,9,9-tetraisopropyl-2,10-dimethyl-4,8-dioxa-3,9-disilaundecan-6-ol, Tetrakis(1-methylethyl)-2,10-dimethyl-3,3,9,9-4,8-dioxa-3,9-disilaundecan-6-ol
IUPAC-namn 1,3-bis[tri(propan-2-yl)silyloxy]propan-2-ol
LEDER CC(C)[Si](OCC(O)CO[Si](C(C)C)(C(C)C)C(C)C)(C(C)C)C(C)C
Specifikationer
Kemiskt namn eller material 1,3-O-Bis(triisopropylsilyl) Glycerol
Kvantitet 25 g
InChI formel InChI=1S/C21H48O3Si2/c1-15(2)25(16(3)4,17(5)6)23-13-21(22)14-24-26(18(7)8,19(9)10)20(11)12/h15-22H,13-14H2,1-12H3
Formel vikt 404.3142