Läs mer
1,3-Propanediyl Bismethanethiosulfonate, TRC
Produktkod. 30420411
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420411 25 mg
30417495 50 mg
30409620 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76035025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55-96-9
Molekylformel C5 H12 O4 S4
Molekylvikt (g/mol) 264.41
IUPAC-namn 1,3-bis(methylsulfonylsulfanyl)propane
LEDER CS(=O)(=O)SCCCSS(=O)(=O)C
Specifikationer
Kemiskt namn eller material 1,3-Propanediyl Bismethanethiosulfonate
Kvantitet 25 mg
InChI formel InChI=1S/C5H12O4S4/c1-12(6,7)10-4-3-5-11-13(2,8)9/h3-5H2,1-2H3
Formel vikt 263.9618