Läs mer
1-[4-(2-hydroxypropan-2-yl)phenyl]ethan-1-one, TRC
Produktkod. 17981445
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981445 5 mg
17922465 50 mg
17992455 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4476505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54549-72-3
Molekylformel C11H14O2
Molekylvikt (g/mol) 178.22
IUPAC-namn 1-[4-(2-hydroxypropan-2-yl)phenyl]ethanone
LEDER CC(=O)C1=CC=C(C=C1)C(C)(C)O
Specifikationer
Kemiskt namn eller material 1-[4-(2-hydroxypropan-2-yl)phenyl]ethan-1-one
Kvantitet 5 mg
InChI formel InChI=1S/C11H14O2/c1-8(12)9-4-6-10(7-5-9)11(2,3)13/h4-7,13H,1-3H3
Formel vikt 178.0994