Läs mer
1-{4-[(2S)-6,15-Dibromo-13-chloro-4-azatricyclo[9.4.0.0]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl]piperidin-1-yl}-2-(piperidin-4-yl)ethan-1-one, TRC
Produktkod. 30364867
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364867 25 mg
30356256 10 mg
30360947 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41491825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1099484-75-9
Molekylformel C26H30N3OClBr2
Molekylvikt (g/mol) 595.8
Synonym (S)-1-(4-(4,8-Dibromo-2-chloro-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-yl)piperidin-1-yl)-2-(piperidin-4-yl)ethan-1-one
IUPAC-namn 1-[4-[(11S)-3,10-dibromo-8-chloro-6,11-dihydro-5H-benzo[1,2]cyclohepta[2,4-b]pyridin-11-yl]piperidin-1-yl]-2-piperidin-4-ylethanone
LEDER Clc1cc(Br)c2[C@H](C3CCN(CC3)C(=O)CC4CCNCC4)c5ncc(Br)cc5CCc2c1
Specifikationer
Kemiskt namn eller material 1-(4-[(2S)-6,15-Dibromo-13-chloro-4-azatricyclo[9.4.0.0]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl]piperidin-1-yl)-2-(piperidin-4-yl)ethan-1-one
Kvantitet 25 mg
InChI formel InChI=1S/C26H30Br2ClN3O/c27-20-12-19-2-1-18-13-21(29)14-22(28)24(18)25(26(19)31-15-20)17-5-9-32(10-6-17)23(33)11-16-3-7-30-8-4-16/h12-17,25,30H,1-11H2/t25-/m0/s1
Formel vikt 593.0444