Läs mer
1-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butyl-7-hydroxy-3,4-dihydro Carbostyril (Aripiprazole Impurity), TRC
Produktkod. 30007290
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007290 1 mg
30005034 50 mg
30004738 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007290 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4361201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797983-65-3
Molekylformel C23 H27 Cl2 N3 O2
Molekylvikt (g/mol) 448.39
Synonym 2(1H)-Quinolinone, 1-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butyl]-3,4-dihydro-7-hydroxy-, 1-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butyl]-7-hydroxy-3,4-dihydroquinolin-2(1H)-one
IUPAC-namn 1-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butyl]-7-hydroxy-3,4-dihydroquinolin-2-one
LEDER Oc1ccc2CCC(=O)N(CCCCN3CCN(CC3)c4cccc(Cl)c4Cl)c2c1
Specifikationer
Kemiskt namn eller material 1-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butyl]-7-hydroxy-3,4-dihydroquinolin-2(1H)-one
Kvantitet 1 mg
InChI formel InChI=1S/C23H27Cl2N3O2/c24-19-4-3-5-20(23(19)25)27-14-12-26(13-15-27)10-1-2-11-28-21-16-18(29)8-6-17(21)7-9-22(28)30/h3-6,8,16,29H,1-2,7,9-15H2
Formel vikt 447.148