Learn More
1-(4-Amino-3-chlorophenyl)ethanone, TRC
Product Code. 30321778
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30321778 1 g
1 options
Product Code. 30321778 Supplier Toronto Research Chemicals Supplier No. TRCA6201581G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1-(4-Amino-3-chlorophenyl)ethanone
CAS 6953-83-9
Molekylformel C8 H8 Cl N O
Kvantitet 1 g
Synonym 1-(4-Amino-3-chlorophenyl)ethan-1-one, 3'-Chloro-4'-aminoacetophenone, 4-Amino-3-chloroacetophenone, NSC 39777
InChI formel InChI=1S/C8H8ClNO/c1-5(11)6-2-3-8(10)7(9)4-6/h2-4H,10H2,1H3
LEDER CC(=O)c1ccc(N)c(Cl)c1
IUPAC-namn 1-(4-amino-3-chlorophenyl)ethanone
Molekylvikt (g/mol) 169.608
Formel vikt 169.029