missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid bis(pinacol) ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7034059
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
99770-93-1
C18H28B2O4
330.038
MFCD08276852
UOJCDDLTVQJPGH-UHFFFAOYSA-N
1,4-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 1,4-benzenediboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-phenylenediboronic acid, pinacol ester, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-benzenediboronic acid dipinacol ester, amtb185, 1,4-phenylenediboronic acid pinacol ester
10592495
4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)B3OC(C(O3)(C)C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
99770-93-1
C18H28B2O4
330.038
MFCD08276852
UOJCDDLTVQJPGH-UHFFFAOYSA-N
1,4-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 1,4-benzenediboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-phenylenediboronic acid, pinacol ester, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-benzenediboronic acid dipinacol ester, amtb185, 1,4-phenylenediboronic acid pinacol ester
10592495
4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)B3OC(C(O3)(C)C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 330.03
Percent Purity 97%
Chemical Name or Material 1,4-Benzenediboronic acid bis(pinacol) ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'