Läs mer
1-(4-Benzyloxy-3-nitrophenyl)-2-bromoethanone, Mikromol™
Produktkod. 30461840
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461840 4 x 25 mg
30051522 100 mg
30052210 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461840 Leverantör Mikromol Leverantörsnummer MM0447.1300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Benzyloxy-3-nitrophenyl)-2-bromoethanone, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(4-Benzyloxy-3-nitrophenyl)-2-bromoethanone
CAS 43229-01-2
Analyt- eller komponentnamn 1-(4-Benzyloxy-3-nitrophenyl)-2-bromoethanone
Frakt skick Room Temperature
Molekylformel C15 H12 Br N O4
Certifikat och efterlevnad ISO 9001
Synonym Fenticonazole Sulfoxide, 2-Bromo-1-[3-nitro-4-(phenylmethoxy)phenyl]ethanone, 1-(Benzyloxy)-2-nitro-4-(2-bromoacetyl)benzene, 2-Bromo-1-(3-nitro-4-benzyloxyphenyl)ethanone, 2-Bromo-4'-Benzyloxy-3'-nitroacetophenone, 4'-(Benzyloxy)-2-bromo-3'-nitroacetophenone
Lösningstyp Neat
LEDER [O-][N+](=O)c1cc(ccc1OCc2ccccc2)C(=O)CBr
Molekylvikt (g/mol) 350.17
Kvantitet 4 x 25 mg
Formel vikt 350.168 g/mol