Läs mer
1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine Dihydrochloride, TRC
Produktkod. 30134532
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134532 5 g
30006209 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134532 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4190605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856841-95-7
Molekylformel C30H30Cl4N2
Molekylvikt (g/mol) 560.38
Synonym USP Cetirizine Dimer Dihydrochloride, Cetririzine EP Impurity D Dihydrochloride;
IUPAC-namn 1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
LEDER CC1=CC=C(C(C2CCC(C(C3=CC=C(C)C=C3)C4=CC=CC=C4)CC2)C5=CC=CC=C5)C=C1
Specifikationer
Kemiskt namn eller material 1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine Dihydrochloride
Kvantitet 5 g
InChI formel InChI=1S/C30H28Cl2N2.2ClH/c31-27-15-11-25(12-16-27)29(23-7-3-1-4-8-23)33-19-21-34(22-20-33)30(24-9-5-2-6-10-24)26-13-17-28(32)18-14-26;;/h1-18,29-30H,19-22H2;2*1H