Läs mer
1,4-Bis(benzhydryl)piperazine, TRC
Produktkod. 17931005
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17931005 1 g
17914695 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17931005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4085001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 216581-01-0
Molekylformel C30 H30 N2
Molekylvikt (g/mol) 418.57
Synonym Piperazine, 1,4-bis(diphenylmethyl)-, 1,4-Bis(diphenylmethyl)piperazine, 1,4-Bisbenzhydrylpiperazine, 1,4-Bis(benzhydryl)piperazine
IUPAC-namn 1,4-dibenzhydrylpiperazine
LEDER C1CN(CCN1C(c2ccccc2)c3ccccc3)C(c4ccccc4)c5ccccc5
Specifikationer
Kemiskt namn eller material 1,4-Bis(diphenylmethyl)piperazine
Kvantitet 1 g
InChI formel InChI=1S/C30H30N2/c1-5-13-25(14-6-1)29(26-15-7-2-8-16-26)31-21-23-32(24-22-31)30(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-20,29-30H,21-24H2
Formel vikt 418.2409