Läs mer
1,4-Bis[(tetrahydrofuran-2-yl)carbonyl]piperazine, Mikromol™
Produktkod. 30049748
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049748 100 mg
30052519 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049748 Leverantör Mikromol Leverantörsnummer MM0843.16
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,4-Bis[(tetrahydrofuran-2-yl)carbonyl]piperazine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,4-Bis[(tetrahydrofuran-2-yl)carbonyl]piperazine
CAS 547730-06-3
Analyt- eller komponentnamn 1,4-Bis[(tetrahydrofuran-2-yl)carbonyl]piperazine
Frakt skick Room Temperature
Molekylformel C14 H22 N2 O4
Certifikat och efterlevnad ISO 17025
Synonym 1,4-Bis[(tetrahydrofuran-2-yl)carbonyl]piperazine, Terazosin Hydrochloride Dihydrate Imp. O (EP), Terazosin Imp. O (EP), Terazosin Hydrochloride Dihydrate Impurity O, Terazosin Impurity O
InChI formel InChI=1S/C14H22N2O4/c17-13(11-3-1-9-19-11)15-5-7-16(8-6-15)14(18)12-4-2-10-20-12/h11-12H,1-10H2
LEDER O=C(C1CCCO1)N2CCN(CC2)C(=O)C3CCCO3
IUPAC-namn [4-(oxolane-2-carbonyl)piperazin-1-yl]-(oxolan-2-yl)methanone
Molekylvikt (g/mol) 282.34
Kvantitet 100 mg
Formel vikt 282.158 g/mol