missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64681.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489066

  • 3249.18 SEK / 1g


Estimated Shipment date
from Supplier 02-08-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
MFCD26960708

Specifications

1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
97%
389.17
1g
MFCD26960708
Videos
SDS